Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RASGEF1B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568295
Description
Predicted to react with Mouse and Rat samples.
Guanine nucleotide exchange factor (GEF) with specificity for RAP2A, it doesn't seems to activate other Ras family proteins (in vitro).
Specifications
RASGEF1B | |
Polyclonal | |
Unconjugated | |
Rasgef1b | |
4732452O09Rik; GPI gamma-4; Gpig4; GPI-gamma 4; GPIgamma4; RasGEF domain family member 1B; RasGEF domain family, member 1B; ras-GEF domain-containing family member 1B; RASGEF1B; RGD1562220 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
100361238, 153020, 320292 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q0VAM2, Q8JZL7 | |
Rasgef1b | |
Synthesized peptide derived from human RASGEF1B. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction