Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RASGRF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559803
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559803 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559803 Supplier Invitrogen™ Supplier No. PA559803
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RKLSLNIPII TGGKALDLAA LSCNSNGYTS MYSAMSPFSK ATLDTSKLYV SSSFTNKIPD EGDTTPEKPE DPSALSKQSS EVSMREESDI DQNQSDDGDT E Highest antigen sequence identity to the following orthologs: Mouse - 58%, Rat - 55%.

RASGRF1 is a guanine nucleotide exchange factor (GEF) similar to the Saccharomyces cerevisiae CDC25 gene product. Functional analysis has demonstrated that this protein stimulates the dissociation of GDP from RAS protein. Studies of the similar gene in mouse suggested that the Ras-GEF activity of this protein in brain can be activated by Ca2+ influx, muscarinic receptors, and G protein beta-gamma subunit. Mouse studies also indicated that the Ras-GEF signaling pathway mediated by this protein may be important for long-term memory.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RASGRF1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RASGRF1
Gene Accession No. Q13972
Gene Alias AI844718; ASPP1; Cdc25; CDC25L; CDC25Mm; Gnrp; GRF1; GRF55; Grfbeta; guanine nucleotide exchange factor; guanine nucleotide-releasing factor 1; guanine nucleotide-releasing factor, 55 kD; guanine nucleotide-releasing protein; H-GRF55; KIAA0771; P140 Ras-GRF; p190; P190-A; p190RhoGEF; PP13187; PPP1R13B; Ras guanine release factor beta; Ras protein specific guanine nucleotide releasing factor 1; RAS protein-specific guanine nucleotide-releasing factor 1; Rasgrf1; ras-GRF1; Ras-specific guanine nucleotide-releasing factor 1; Ras-specific guanine nucleotide-releasing factor, CDC25 homolog; ras-specific nucleotide exchange factor CDC25
Gene Symbols RASGRF1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RASGRF1. Recombinant protein control fragment (Product #RP-98034).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5923
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.