Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RASGRF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578412
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578412 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578412 Supplier Invitrogen™ Supplier No. PA578412
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: mouse brain, rat brain Predicted reactivity: Rat (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

RASGRF1 is a guanine nucleotide exchange factor (GEF) similar to the Saccharomyces cerevisiae CDC25 gene product. Functional analysis has demonstrated that this protein stimulates the dissociation of GDP from RAS protein. Studies of the similar gene in mouse suggested that the Ras-GEF activity of this protein in brain can be activated by Ca2+ influx, muscarinic receptors, and G protein beta-gamma subunit. Mouse studies also indicated that the Ras-GEF signaling pathway mediated by this protein may be important for long-term memory.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RASGRF1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene RASGRF1
Gene Accession No. P27671, P28818
Gene Alias AI844718; ASPP1; Cdc25; CDC25L; CDC25Mm; Gnrp; GRF1; GRF55; Grfbeta; guanine nucleotide exchange factor; guanine nucleotide-releasing factor 1; guanine nucleotide-releasing factor, 55 kD; guanine nucleotide-releasing protein; H-GRF55; KIAA0771; P140 Ras-GRF; p190; P190-A; p190RhoGEF; PP13187; PPP1R13B; Ras guanine release factor beta; Ras protein specific guanine nucleotide releasing factor 1; RAS protein-specific guanine nucleotide-releasing factor 1; Rasgrf1; ras-GRF1; Ras-specific guanine nucleotide-releasing factor 1; Ras-specific guanine nucleotide-releasing factor, CDC25 homolog; ras-specific nucleotide exchange factor CDC25
Gene Symbols RASGRF1
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of Rat RASGRF1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 192213, 19417
Target Species Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.