Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RASGRF2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553842
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553842 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553842 Supplier Invitrogen™ Supplier No. PA553842
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FATSQNNRGE HLVDGKSPRL CRKFSSPPPL AVSRTSSPVR ARKLSLTSPL NSKIGALDLT TSSSPTTTTQ SPAASPPPHT GQIPLDLSRG LSSPEQSPGT VEENVD Highest antigen sequence identity to the following orthologs: Mouse - 72%, Rat - 71%.

RAS (MIM 190020) GTPases cycle between an inactive GDP-bound state and an active GTP-bound state. Guanine-nucleotide exchange factors (GEFs), such as RASGRFs, stimulate the conversion of the GDP-bound form into the active form.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RASGRF2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RASGRF2
Gene Accession No. O14827
Gene Alias 6330417G04Rik; AW048350; Grf2; guanine nucleotide-releasing factor 2; ras guanine nucleotide exchange factor 2; Ras protein specific guanine nucleotide releasing factor 2; RAS protein-specific guanine nucleotide-releasing factor 2; Rasgrf2; ras-GRF2; Ras-specific guanine nucleotide-releasing factor 2
Gene Symbols RASGRF2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RASGRF2. Recombinant protein control fragment (Product #RP-92409).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5924
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.