Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RASGRP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587944
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587944 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587944 Supplier Invitrogen™ Supplier No. PA587944
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PVAPTENNTS VGPVSNLCSL GAKDLLHAPE EGPFTFPNGE AVEHGEESKD RTIMLMGVSS QKISLRLKRA VAHKATQTES QPWIGSEGPS GPFVLSSPRK TAQDTLYVLP SPTSPCPSPV LVRKRAFVKW ENKDSLIKSK EELRHLRLPT YQELEQEINT LKADNDALKI QLKYAQKKIE SLQLEKSNHV LAQMEQGDCS; Positive Samples: Jurkat, MCF7, A-549, U-251MG, SW480; Cellular Location: Cell membrane, Cytoplasm, Endoplasmic reticulum membrane, Golgi apparatus membrane, Peripheral membrane protein, cytosol.

Functions as a calcium- and diacylglycerol (DAG)-regulated nucleotide exchange factor specifically activating Ras through the exchange of bound GDP for GTP. Activates the Erk/MAP kinase cascade. Regulates T-cell/B-cell development, homeostasis and differentiation by coupling T-lymphocyte/B-lymphocyte antigen receptors to Ras. Regulates NK cell cytotoxicity and ITAM-dependent cytokine production by activation of Ras-mediated ERK and JNK pathways. Functions in mast cell degranulation and cytokine secretion, regulating FcERI-evoked allergic responses. May also function in differentiation of other cell types.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RASGRP1
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.99 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene RASGRP1
Gene Accession No. O95267, Q9R1K8, Q9Z1S3
Gene Alias Calcium and DAG-regulated guanine nucleotide exchange factor II; calcium- and diacylglycerol-regulated guanine nucleotide exchange factor II; CALDAG-GEFI; CALDAG-GEFII; guanine nucleotide exchange factor, calcium- and DAG-regulated, Rap1A; hRasGRP1; ras activator RasGRP; RAS guanyl nucleotide-releasing protein 1; RAS guanyl releasing protein 1; RAS guanyl releasing protein 1 (calcium and DAG-regulated); Ras guanyl-releasing protein; RAS guanyl-releasing protein 1; RASGRP; RASGRP1; V
Gene Symbols RASGRP1
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10125, 19419, 29434
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.