Learn More
Invitrogen™ RASGRP1 Polyclonal Antibody

Description
Sequence homology: Cow: 86%; Dog: 93%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 79%; Rabbit: 79%; Rat: 93%.
Specifications
Specifications
| Antigen | RASGRP1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 |
| Gene | RASGRP1 |
| Gene Accession No. | O95267 |
| Gene Alias | Calcium and DAG-regulated guanine nucleotide exchange factor II; calcium- and diacylglycerol-regulated guanine nucleotide exchange factor II; CALDAG-GEFI; CALDAG-GEFII; guanine nucleotide exchange factor, calcium- and DAG-regulated, Rap1A; hRasGRP1; ras activator RasGRP; RAS guanyl nucleotide-releasing protein 1; RAS guanyl releasing protein 1; RAS guanyl releasing protein 1 (calcium and DAG-regulated); Ras guanyl-releasing protein; RAS guanyl-releasing protein 1; RASGRP; RASGRP1; V |
| Gene Symbols | RASGRP1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.