Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RASGRP2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP1588720 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP1588720 20 μL
NBP158871 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP1588720 Supplier Novus Biologicals Supplier No. NBP15887120UL

Rabbit Polyclonal Antibody

RASGRP2 Polyclonal specifically detects RASGRP2 in Human samples. It is validated for Western Blot.

Specifications

Antigen RASGRP2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q7LDG7
Gene Alias Calcium and DAG-regulated guanine nucleotide exchange factor I, CALDAG-GEFI, CDC25Lcalcium and diacylglycerol-regulated guanine nucleotide exchange factor I, Cdc25-like protein, F25B3.3 kinase-like protein, guanine exchange factor MCG7, hCDC25L, MCG7, RAS guanyl nucleotide-releasing protein 2, RAS guanyl releasing protein 2 (calcium and DAG-regulated), RAS guanyl-releasing protein 2
Gene Symbols RASGRP2
Host Species Rabbit
Immunogen Synthetic peptides corresponding to RASGRP2(RAS guanyl releasing protein 2 (calcium and DAG-regulated)) The peptide sequence was selected from the N terminal of RASGRP2. Peptide sequence LVRMFLMMHPWYIPSSQLAAKLLHIYQQSRKDNSNSLQVKTCHLVRYWIS.
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10235
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.