Learn More
Invitrogen™ RASGRP3 Monoclonal Antibody (OTI2C11)

Description
RASGRP3 Monoclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | RASGRP3 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI2C11 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | Rasgrp3 |
| Gene Accession No. | Q8IV61 |
| Gene Alias | BC066069; calcium and DAG-regulated guanine nucleotide exchange factor III; calcium- and diacylglycerol-regulated guanine nucleotide exchange factor III; CalDAG-GEFIII; Gm327; GRP3; Guanine nucleotide exchange factor for Rap1; KIAA0846; OTTHUMP00000201245; RAS guanyl releasing protein 3; RAS guanyl releasing protein 3 (calcium and DAG-regulated); ras guanyl-releasing protein 3; RAS, guanyl releasing protein 3; Rasgrp3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.