Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RASGRP3 Monoclonal Antibody (OTI2C9)
GREENER_CHOICE

Catalog No. PIMA526499
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526499 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526499 Supplier Invitrogen™ Supplier No. MA526499
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RASGRP3 Monoclonal Antibody for Western Blot, IHC (P)

Members of the RAS subfamily of GTPases function in signal transduction as GTP/GDP-regulated switches that cycle between inactive GDP- and active GTP-bound states. Guanine nucleotide exchange factors (GEFs), such as RASGRP3, serve as RAS activators by promoting acquisition of GTP to maintain the active GTP-bound state and are the key link between cell surface receptors and RAS activation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RASGRP3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI2C9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Rasgrp3
Gene Accession No. Q8IV61
Gene Alias BC066069; calcium and DAG-regulated guanine nucleotide exchange factor III; calcium- and diacylglycerol-regulated guanine nucleotide exchange factor III; CalDAG-GEFIII; Gm327; GRP3; Guanine nucleotide exchange factor for Rap1; KIAA0846; OTTHUMP00000201245; RAS guanyl releasing protein 3; RAS guanyl releasing protein 3 (calcium and DAG-regulated); ras guanyl-releasing protein 3; RAS, guanyl releasing protein 3; Rasgrp3
Gene Symbols Rasgrp3
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 380-690 of RASGRP3 produced in E.coli.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25780
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.