Learn More
RASGRP3 Mouse anti-Human, Clone: OTI5D2, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF801532

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Members of the RAS subfamily of GTPases function in signal transduction as GTP/GDP-regulated switches that cycle between inactive GDP- and active GTP-bound states. Guanine nucleotide exchange factors (GEFs), such as RASGRP3, serve as RAS activators by promoting acquisition of GTP to maintain the active GTP-bound state and are the key link between cell surface receptors and RAS activation.Specifications
RASGRP3 | |
Monoclonal | |
Unconjugated | |
RASGRP3 | |
GRP3 | |
Mouse | |
Affinity Chromatography | |
RUO | |
25780 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI5D2 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q8IV61 | |
RASGRP3 | |
Human recombit protein fragment corresponding to amino acids 380-690 of human RASGRP3 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.