Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RASGRP3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566222
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566222 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566222 Supplier Invitrogen™ Supplier No. PA566222
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RYWILKFPAEF NLDLGLIRMT EEFREVASQL GYEKHVSLID ISSIPSYDWM RRVTQRKKVS KKGKACLLFD HLE Highest antigen sequence identity to the following orthologs - mouse 93%, rat 93%.

Members of the RAS subfamily of GTPases function in signal transduction as GTP/GDP-regulated switches that cycle between inactive GDP- and active GTP-bound states. Guanine nucleotide exchange factors (GEFs), such as RASGRP3, serve as RAS activators by promoting acquisition of GTP to maintain the active GTP-bound state and are the key link between cell surface receptors and RAS activation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RASGRP3
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Rasgrp3
Gene Accession No. Q8IV61
Gene Alias BC066069; calcium and DAG-regulated guanine nucleotide exchange factor III; calcium- and diacylglycerol-regulated guanine nucleotide exchange factor III; CalDAG-GEFIII; Gm327; GRP3; Guanine nucleotide exchange factor for Rap1; KIAA0846; OTTHUMP00000201245; RAS guanyl releasing protein 3; RAS guanyl releasing protein 3 (calcium and DAG-regulated); ras guanyl-releasing protein 3; RAS, guanyl releasing protein 3; Rasgrp3
Gene Symbols Rasgrp3
Host Species Rabbit
Immunogen Recombinant Human RASGRP3. Recombinant protein control fragment (Product #RP-103934).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25780
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.