Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RASIP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116841
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116841 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116841 Supplier Invitrogen™ Supplier No. PA5116841
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: Mouse skeletal muscle Immunogen sequence: TPEAVSVELR PLMLWMANTT ELLSFVQEKV LEMEKEADQE DPQLCNDLEL CDEAMALLDE VIMCTFQQSV YYLTKTLYST LPALLDSNPF TAGAELPGPG AELGAMPPGL RPTLGVFQAA LELTSQCELH PDLVSQTFGY LFFFSNASLL N.

Required for the proper formation of vascular structures that develop via both vasculogenesis and angiogenesis. Acts as a critical and vascular-specific regulator of GTPase signaling, cell architecture, and adhesion, which is essential for endothelial cell morphogenesis and blood vessel tubulogenesis. Regulates the activity of Rho GTPases in part by recruiting ARHGAP29 and suppressing RhoA signaling and dampening ROCK and MYH9 activities in endothelial cells. May act as effector for Golgi-bound HRAS and other Ras-like proteins. May promote HRAS-mediated transformation. Negative regulator of amino acid starvation-induced autophagy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RASIP1
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.04 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene RASIP1
Gene Accession No. Q3U0S6, Q5U651
Gene Alias 1110025D03Rik; 2610025P08Rik; AI853551; RAIN; Ras interacting protein 1; ras-interacting protein 1; Rasip1
Gene Symbols RASIP1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 635-785 of human RASIP1 (Q5U651).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 292912, 54922, 69903
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.