Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RASL10A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP234210
Description
RASL10A Polyclonal specifically detects RASL10A in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
RASL10A | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
Q92737 | |
RASL10A | |
This antibody was developed against a recombinant protein corresponding to amino acids: AKYNWHVLRLFRELLRCALVRARPAHPALRLQGALHPARCSLM | |
0.1 mL | |
Signal Transduction | |
10633 | |
Human | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
ras-like protein family member 10A, RAS-like, family 10, member A, RAS-related on chromosome 22, Ras-related protein on chromosome 22, RRP22Ras-like protein RRP22 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction