Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RASL10A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA551054
Description
RASL10A Polyclonal Antibody detects endogenous levels of RASL10A protein.
The specific function of RASL10A is not yet known.
Specifications
RASL10A | |
Polyclonal | |
Unconjugated | |
RASL10A | |
2210403B10Rik; AI852688; RAS like family 10 member A; RASL10A; RAS-like family 10 member A; ras-like protein family member 10A; Ras-like protein RRP22; Ras-like protein RRP22 (RAS-related protein on chromosome 22); RAS-like, family 10, member A; ras-related protein on chromosome 22; Ras-related protein on chromosome 22 homolog; RGD1306100; RRP22 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10633, 75668 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1 mg/mL | |
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q8K5A4, Q92737 | |
RASL10A | |
Synthesized peptide derived from the Internal region of human RRP22. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction