Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RASL10A Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986750
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6750 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6750 Supplier Bioss Supplier No. BS5722R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The specific function of RASL10A is not yet known.

Specifications

Antigen RASL10A
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene RASL10A
Gene Accession No. Q92737
Gene Alias 2210403B10Rik; AI852688; RAS like family 10 member A; Rasl10a; RAS-like family 10 member A; Ras-like protein family member 10A; Ras-like protein RRP22; Ras-like protein RRP22 (RAS-related protein on chromosome 22); RAS-like, family 10, member A; Ras-related protein on chromosome 22; Ras-related protein on chromosome 22 homolog; Rrp22
Gene Symbols RASL10A
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human RASL.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10633
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.