Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RASL10B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563398
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563398 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563398 Supplier Invitrogen™ Supplier No. PA563398
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KSAIVRQFLY NEFSEVCVPT TARRLYLPAV VMNGHVHDLQ ILDFPPISAF PVN Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 100%.

May facilitate the release of atrial natriuretic peptide by cardiomyocytes and hence play a role in the regulation of arterial pressure.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RASL10B
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RASL10B
Gene Accession No. Q96S79
Gene Alias alternative protein RASL10B; B230331P10Rik; Gm729; RAS like family 10 member B; Rasl10b; RAS-like family 10 member B; ras-like protein family member 10B; Ras-like protein VTS58635; RAS-like, family 10, member B; Ras-related protein 17; RRP17; rslab; VTS58635
Gene Symbols RASL10B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RASL10B. Recombinant protein control fragment (Product #RP-103289).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91608
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.