Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RASL11A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552124
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552124 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552124 Supplier Invitrogen™ Supplier No. PA552124
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VGKSAMIVRF LTKRFIGDYE PNTGKLYSRL VYVEGDQLSL QIQDTPGGVQ IQDSLPQVVD SLSKCVQWAE GFLLVYSITD YDSYLSIRPL YQHIRKVHPD SKAPVIIVGN KGDLLHARQV QTQDGIQ Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 91%.

Regulator of rDNA transcription. Acts in cooperation UBF/UBTF and positively regulates RNA polymerase I transcription.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RASL11A
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RASL11A
Gene Accession No. Q6T310
Gene Alias RAS like family 11 member A; RASL11A; RAS-like family 11 member A; ras-like protein family member 11A; RAS-like, family 11, member A
Gene Symbols RASL11A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RASL11A. Recombinant protein control fragment (Product #RP-101009).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 387496
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.