Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RASL12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570262
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570262 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570262 Supplier Invitrogen™ Supplier No. PA570262
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 93%; Rat: 100%.

RASL12 belongs to the small GTPase superfamily. Ras family.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RASL12
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene RASL12
Gene Accession No. Q08AT1
Gene Alias 4631404I11Rik; LOC100714151; Ras family member Ris; RAS like family 12; Rasl12; RAS-like family 12; ras-like protein family member 12; Ras-like protein Ris; RAS-like, family 12; RAS-like, family 12 protein; RIS
Gene Symbols RASL12
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human RASL12 (aa 50-99).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 70784
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.