Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RASSF1 Antibody (SR1658), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32178450UL
Description
RASSF1 Monoclonal antibody specifically detects RASSF1 in Human samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
| RASSF1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry | |
| SR1658 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
| 123F2, NORE2A, pancreas-specific ras association domain family 1 protein, Ras association (RalGDS/AF-6) domain family member 1, ras association domain-containing protein 1, RASSF1A, RDA32cardiac-specific ras association domain family 1 protein, REH3P21, tumor suppressor protein RDA32, WUGSC:H_LUCA12.5 | |
| A synthesized peptide derived from human RASSF1 (Uniprot #: Q9NS23) | |
| 50 μg | |
| Breast Cancer, Cell Cycle and Replication, Mitotic Regulators | |
| 11186 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction