Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RASSF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588305
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588305 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588305 Supplier Invitrogen™ Supplier No. PA588305
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CCGPRDLGWE PAVERDTNVD EPVEWETPDL SQAEIEQKIK EYNAQINSNL FMSLNKDGSY TGFIKVQLKL VRPVSVPSSK KPPSLQDARR GPGRGTSVRR RTSFYLPKDA VKHLHVLSRT RAREVIEALL RKFLVVDDPR KFALFERAER HGQVYLRKLL DDEQPLRLRL LAGPSDKALS FVLKENDSGE VNWDAFSMPE LHNFLRILQR EEEEHLRQIL QKYSYCRQKI QEALHACPLG; Positive Samples: HeLa, HepG2, 293T, SW480; Cellular Location: Cytoplasm, Nucleus, Nucleus, centrosome, cytoskeleton, microtubule organizing center, spindle, spindle pole.

This gene encodes a protein similar to the RAS effector proteins. Loss or altered expression of this gene has been associated with the pathogenesis of a variety of cancers, which suggests the tumor suppressor function of this gene. The inactivation of this gene was found to be correlated with the hypermethylation of its CpG-island promoter region. The encoded protein was found to interact with DNA repair protein XPA. The protein was also shown to inhibit the accumulation of cyclin D1, and thus induce cell cycle arrest. Seven alternatively spliced transcript variants of this gene encoding distinct isoforms have been reported.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RASSF1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.68 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene RASSF1
Gene Accession No. Q9NS23
Gene Alias 123F protein; 123F2; AA536941; AU044980; cardiac-specific ras association domain family 1 protein; NORE2A; pancreas-specific ras association domain family 1 protein; protein 123F2; Ras association (RalGDS/AF-6) domain family 1; Ras association (RalGDS/AF-6) domain family member 1; Ras association domain family 1; Ras association domain family member 1; ras association domain-containing protein 1; Rassf1; Rassf1A; RASSF1A tumor suppressor; Rassf1B; Rassf1C; RDA32; REH3P21; tumor suppressor protein RDA32; wu:fc17f02; WUGSC:H_LUCA12.5; zgc:112224; zgc:92505; zgc:92505 protein
Gene Symbols RASSF1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 101-340 of human RASSF1 (NP_0091133).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11186
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.