Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RASSF1, Rabbit anti-Rat, Human, Polyclonal, Millipore Sigma™

Catalog No. ABS148425UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABS148425UL 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABS148425UL Supplier MilliporeSigma Supplier No. ABS148425UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-RASSF1, Cat. No. ABS1484, is a rabbit polyclonal antibody tht detects Ras association domain-containing protein 1 in human and rat tissues and has been tested for use in Flow Cytometry, Immunohistochemistry (paraffin), and Western Blotting.

Ras association domain-containing protein 1 (UniProt: Q9NS23; RASSF1) is encoded by the RASSF1 (also known as RDA32) gene (Gene ID: 11186) in human. RASSF1 serves as a potential tumor suppressor and is shown to be essential for death receptor-dependent apoptosis. It mediates the activation of STK3/MST2 and STK4/MST1 during Fas-induced apoptosis by preventing their dephosphorylation. In association with MOAP1, it promotes Bax conformational change and translocation to mitochondrial membranes in response to TNF and TNFSF10 stimulation. Eight isoforms of RASSF1 have been described that are produced by alternative promoter usage and alternative splicing. The immunogen sequence used to generate this antibody is conserved in isoforms A, D, E, F and G. Isoform A is ubiquitously expressed and is shown to localize to cytoplasmic microtubules during interphase, to bipolar centrosomes associated with microtubules during prophase, to spindle fibers and spindle poles at metaphase and anaphase, to the midzone during early telophase, and to the midbody in late telophase in cells. Isoform A is reported to interact with CDC20, an activator of the anaphase-promoting complex (APC), resulting in the inhibition of APC activity and mitotic progression. It Inhibits proliferation by negatively regulating cell cycle progression at the level of G1/S-phase transition by regulating accumulation of cyclin D1 protein. It also disrupts interactions among MDM2, DAXX and USP7 and thereby contributes to the efficient activation of TP53 by promoting MDM2 self-ubiquitination in cell-cycle checkpoint control in response to DNA damage.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RASSF1
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Flow Cytometry Analysis: 1μg from a representative lot detected RASSF1 in Jurkat cells.Immunohistochemistry Analysis: A 1:250 dilution from a representative lot detected RASSF1 in human ovarian cancer, human placenta, and human pancreas tissue sections.
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Ras association domain-containing protein 1
Gene Symbols RASSF1;RDA32
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to 14 amino acids from the N-terminal region of human Ras association domain-containing protein 1.
Molecular Weight of Antigen ∽39 kDa observed; 39.22 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Test Specificity This rabbit polyclonal antibody detects Ras association domain-containing protein 1 in human and rat tissues. It targets an epitope within 14 amino acids from the N-terminal region.
Target Species Rat, Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.