Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RASSF10 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP2411300.025MG
Description
RASSF10 Polyclonal specifically detects RASSF10 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
RASSF10 | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
RASSF10 | |
Antibody was raised against a 17 amino acid synthetic peptide near the amino terminus of human RASSF10. The immunogen is located within amino acids 50 - 100 of RASSF10. Amino Acid Squence: HGPGELPEPPNEDDEDD | |
0.025 mg | |
Primary | |
RASSF10 antibody will not cross-react with other RAS association domain-containing family members. | |
Store at 4°C. |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
Western Blot 1 μg/mL ELISA, Immunohistochemistry 5 μg/mL, Immunohistochemistry-Paraffin 5 μg/mL | |
Peptidylglycine Alpha-Amidating Monooxygenase COOH-Terminal Interactor-Like, Ras Association (RalGDS/AF-6) Domain Family (N-Terminal) Member 10, Ras Association Domain-Containing Protein 10 | |
Rabbit | |
Affinity Purified | |
RUO | |
644943 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction