Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RASSF10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100198
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100198 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100198 Supplier Thermo Scientific Supplier No. PA5100198
Add to Cart
Add to Cart

RASSF10 Polyclonal Antibody

RASSF10 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen RASSF10
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene RASSF10
Gene Accession No. A6NK89
Gene Alias 4632411J06Rik; Ras association (RalGDS/AF-6) domain family (N-terminal) member 10; Ras association domain family member 10; ras association domain-containing protein 10; Rassf10
Gene Symbols RASSF10
Host Species Rabbit
Immunogen A synthesized peptide derived from human RASSF10(Accession A6NK89), corresponding to amino acid residues M466-V507.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 644943
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.