Learn More
Invitrogen™ RASSF10 Polyclonal Antibody

Description
A suggested positive control is human lung tissue lysate. PA5-21151 can be used with blocking peptide PEP-1265.
Specifications
Specifications
| Antigen | RASSF10 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | RASSF10 |
| Gene Accession No. | A6NK89 |
| Gene Alias | 4632411J06Rik; Ras association (RalGDS/AF-6) domain family (N-terminal) member 10; Ras association domain family member 10; ras association domain-containing protein 10; Rassf10 |
| Gene Symbols | RASSF10 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.