Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RASSF2 Monoclonal Antibody (1509CT269.10.9.29)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA524746

Catalog No. PIMA524746


Only null left
Add to Cart

Description

Description

RASSF2 Monoclonal Antibody for Western Blot, Flow

Potential tumor suppressor. Acts as a KRAS-specific effector protein. May promote apoptosis and cell cycle arrest. Stabilizes STK3/MST2 by protecting it from proteasomal degradation.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

RASSF2
Monoclonal
1.46 mg/mL
PBS with 0.09% sodium azide; pH 7.4
P50749
RASSF2
Recombinant protein between 1-326 amino acids from human RASSF2.
200 μL
Primary
Human
Antibody
IgG2a κ
Flow Cytometry, Western Blot
1509CT269.10.9.29
Unconjugated
RASSF2
3830431H01Rik; 9030412M04Rik; AI852669; AW495050; CENP-34; centromere protein 34; Kiaa0168; Ras association (RalGDS/AF-6) domain family 2; Ras association (RalGDS/AF-6) domain family member 2; Ras association domain family member 2; ras association domain-containing protein 2; RASFADIN; Rassf2; RP4-599I11.1
Mouse
Protein G
RUO
9770
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.