Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RASSF2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598113
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598113 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598113 Supplier Thermo Scientific Supplier No. PA598113
Add to Cart
Add to Cart

RASSF2 Polyclonal Antibody

RASSF2 Polyclonal Antibody for Western Blot, IP, ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen RASSF2
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1.83 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene RASSF2
Gene Accession No. P50749
Gene Alias 3830431H01Rik; 9030412M04Rik; AI852669; AW495050; CENP-34; centromere protein 34; Kiaa0168; Ras association (RalGDS/AF-6) domain family 2; Ras association (RalGDS/AF-6) domain family member 2; Ras association domain family member 2; ras association domain-containing protein 2; RASFADIN; Rassf2; RP4-599I11.1
Gene Symbols RASSF2
Host Species Rabbit
Immunogen Recombinant Human Ras association domain-containing protein 2 protein (69-326AA).
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9770
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.