Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RASSF2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103099
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103099 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103099 Supplier Invitrogen™ Supplier No. PA5103099
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total RASSF2.

Potential tumor suppressor. Acts as a KRAS-specific effector protein. May promote apoptosis and cell cycle arrest. Stabilizes STK3/MST2 by protecting it from proteasomal degradation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RASSF2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene RASSF2
Gene Accession No. P50749, Q3B7D5, Q8BMS9
Gene Alias 3830431H01Rik; 9030412M04Rik; AI852669; AW495050; CENP-34; centromere protein 34; Kiaa0168; Ras association (RalGDS/AF-6) domain family 2; Ras association (RalGDS/AF-6) domain family member 2; Ras association domain family member 2; ras association domain-containing protein 2; RASFADIN; Rassf2; RP4-599I11.1
Gene Symbols RASSF2
Host Species Rabbit
Immunogen A synthesized peptide derived from human RASSF2(Accession P50749), corresponding to amino acid residues V112-R162.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 215653, 311437, 9770
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.