Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RASSF3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558406
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558406 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558406 Supplier Invitrogen™ Supplier No. PA558406
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSSGYSSLEE DAEDFFFTAR TSFFRRAPQG KPRSGQQDVE KEKETHSYLS KEEIKEKVHK YNLAVTDKLK MTLNSNGIYT G Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 95%.

The RAS oncogene is mutated in nearly one-third of all human cancers. Members of the RAS superfamily are plasma membrane GTP-binding proteins that modulate intracellular signal transduction pathways. A subfamily of RAS effectors, including RASSF3, share a RAS association domain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RASSF3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RASSF3
Gene Accession No. Q86WH2
Gene Alias AW212023; AW322379; MGC119194; MGC119195; MGC119197; novel ras effector 1; Ras association (RalGDS/AF-6) domain family 3; Ras association (RalGDS/AF-6) domain family member 3; Ras association domain family 3; Ras association domain family member 3; ras association domain-containing protein 3; Rassf3; RASSF5
Gene Symbols RASSF3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RASSF3. Recombinant protein control fragment (Product #RP-97777).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283349
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.