Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RASSF4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558556
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558556 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558556 Supplier Invitrogen™ Supplier No. PA558556
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QDDREQVHLP STSWMPRRPS CPLKEPSPQN GNITAQGPSI QPVHKAESST DSSGPLEEAE EAPQLMRTKS DASCMSQRRP KCRAPGEAQR I Highest antigen sequence identity to the following orthologs: Mouse - 63%, Rat - 64%.

The function of this gene has not yet been determined but may involve a role in tumor suppression. Alternative splicing of this gene results in several transcript variants; however, most of the variants have not been fully described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RASSF4
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RASSF4
Gene Accession No. Q9H2L5
Gene Alias 3830411C14Rik; AD037; AI586137; Ras association (RalGDS/AF-6) domain family 4; Ras association (RalGDS/AF-6) domain family member 4; Ras association domain family 4; Ras association domain family member 4; ras association domain-containing protein 4; Rassf4; tumor suppressor RASSF4
Gene Symbols RASSF4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RASSF4. Recombinant protein control fragment (Product #RP-97020).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83937
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.