Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RASSF5 Monoclonal Antibody (OTI1H2), TrueMAB™, OriGene
SDP

Catalog No. 502541499
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-1499 30 μL
50-167-3562 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-1499 Supplier OriGene Supplier No. TA502509S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RASSF5 Monoclonal antibody specifically detects RASSF5 in Human samples. It is validated for Flow Cytometry, Western Blot

Specifications

Antigen RASSF5
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone OTI1H2
Concentration 0.41 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene RASSF5
Gene Accession No. Q8WWW0
Gene Alias 1300019G20Rik; AU042887; Maxp1; MGC10823; MGC17344; new ras effector 1; NORE1; Nore1A; NORE1B; novel ras effector 1; protein interacting with guanine nucleotide exchange factor; Rap1-binding protein; RAPL; Ras association (RalGDS/AF-6) domain family 5; Ras association (RalGDS/AF-6) domain family member 5; Ras association domain family member 5; ras association domain-containing protein 5; Ras effector-like protein; RASSF3; RASSF5; regulator for cell adhesion and polarization enriched in lymphoid tissue; regulator for cell adhesion and polarization enriched in lymphoid tissues; RP11-343H5.1; tumor; tumor suppressor RASSF3
Host Species Mouse
Immunogen Full length human recombinant protein of human RASSF5 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83593
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.