Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RASSF6 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP281798
Description
RASSF6 Polyclonal specifically detects RASSF6 in Human samples. It is validated for Western Blot, ELISA.Specifications
RASSF6 | |
Polyclonal | |
Unconjugated | |
PBS | |
RASSF6 | |
RASSF6 antibody was raised against a 14 amino acid synthetic peptide near the center of human RASSF6. The immunogen is located within amino acids 230 - 280 of RASSF6. Amino Acid Squence: LQKFKIENSPQDFA | |
0.1 mg | |
Primary | |
At least four isoforms of RASSF6 are known to exist. RASSF6 antibody is predicted to not cross-react with other RASSF family members. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot, ELISA | |
1 mg/mL | |
Western Blot 1 μg/mL, ELISA | |
DKFZp686K23225, putative RAS binding protein, Ras association (RalGDS/AF-6) domain family 6, Ras association (RalGDS/AF-6) domain family member 6, ras association domain-containing protein 6 | |
Rabbit | |
Affinity Purified | |
RUO | |
166824 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction