Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RASSF6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565191
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565191 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565191 Supplier Invitrogen™ Supplier No. PA565191
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SDLPYRISSDH LKKEEKMTMM AHQYPSWIFI NEKTFITREQ LNSLLKTYNI FYENQKNLHI LYGETEDGKL IVEGMLDIFW GVKRPIQ Highest antigen sequence identity to the following orthologs - mouse 66%, rat 67%.

RASSF6 is a member of the RAS association domain family of RAS effectors/tumor suppressors that mediate some of the growth inhibitory properties of RAS oncogenes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RASSF6
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Rassf6
Gene Accession No. Q6ZTQ3
Gene Alias 1600016B17Rik; DKFZp686K23225; putative RAS binding protein; Ras association (RalGDS/AF-6) domain family 6; Ras association (RalGDS/AF-6) domain family member 6; Ras association domain family member 6; ras association domain-containing protein 6; RASSF6
Gene Symbols Rassf6
Host Species Rabbit
Immunogen Recombinant Human RASSF6. Recombinant protein control fragment (Product #RP-104678).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 166824
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.