Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RASSF6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA576480
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA576480 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA576480 Supplier Invitrogen™ Supplier No. PA576480
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

RASSF6 is a member of the RAS association domain family of RAS effectors/tumor suppressors that mediate some of the growth inhibitory properties of RAS oncogenes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RASSF6
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene Rassf6
Gene Accession No. Q4QR82, Q6ZTQ3, Q80UQ2
Gene Alias 1600016B17Rik; DKFZp686K23225; putative RAS binding protein; Ras association (RalGDS/AF-6) domain family 6; Ras association (RalGDS/AF-6) domain family member 6; Ras association domain family member 6; ras association domain-containing protein 6; RASSF6
Gene Symbols Rassf6
Host Species Rabbit
Immunogen Recombinant fusion protein of human RASSF6.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 166824, 305251, 73246
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.