Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RASSF7 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP215091
Description
RASSF7 Polyclonal specifically detects RASSF7 in Human samples. It is validated for Western Blot.Specifications
RASSF7 | |
Polyclonal | |
Unconjugated | |
PBS (pH 7.0) and 20% Glycerol with 0.01% Thimerosal | |
chromosome 11 open reading frame 13, HRAS1-related cluster protein 1, HRAS1-related cluster-1, HRC1C11orf13HRAS1, MGC126069, MGC126070, Ras association (RalGDS/AF-6) domain family (N-terminal) member 7, Ras association (RalGDS/AF-6) domain family 7, ras association domain-containing protein 7 | |
Rabbit | |
40 kDa | |
0.1 mL | |
Primary | |
Human | |
IgG |
Western Blot | |
1mg/mL | |
Western Blot 1:500-1:3000 | |
Q02833 | |
RASSF7 | |
Recombinant protein encompassing a sequence within the center region of human RASSF7. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
8045 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction