Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RASSF7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114128
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114128 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114128 Supplier Invitrogen™ Supplier No. PA5114128
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DISIPDYCSL GDGEEEEITI NAWFGPQGTI SPLHQDPQQN FLVQVMGRKY For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 79%; Dog: 93%; Horse: 77%; Human: 100%; Pig: 79%; Rat: 86%.

Negatively regulates stress-induced JNK activation and apoptosis by promoting MAP2K7 phosphorylation and inhibiting its ability to activate JNK. Following prolonged stress, anti-apoptotic effect stops because of degradation of RASSF7 protein via the ubiquitin-proteasome pathway. Required for the activation of AURKB and chromosomal congression during mitosis where it stimulates microtubule polymerization. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen RASSF7
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene RASSF7
Gene Accession No. Q02833
Gene Alias 2400009B11Rik; AW210608; C11orf13; HRAS1; HRAS1-related cluster protein 1; HRAS1-related cluster-1; HRC1; LOW QUALITY PROTEIN: ras association domain-containing protein 7; MGC126069; MGC126070; Ras association (RalGDS/AF-6) domain family (N-terminal) member 7; Ras association (RalGDS/AF-6) domain family 7; Ras association domain family member 7; ras association domain-containing protein 7; Rassf7; RGD1306244
Gene Symbols RASSF7
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human RASSF7.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8045
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.