Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RASSF8 Antibody (OTI1F5), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273820V
Description
RASSF8 Monoclonal specifically detects RASSF8 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
RASSF8 | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
RASSF8 | |
Full length recombinant protein of human RASSF8 (NP_009142) produced in HEK293T cells. | |
0.1 mL | |
11228.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI1F5 | |
Western Blot, Immunohistochemistry | |
C12orf2, carcinoma associated HOJ-1, Carcinoma-associated protein HOJ-1, chromosome 12 open reading frame 2, DKFZp434O0227, FLJ11542, HoJ-1, HOJ1, Ras association (RalGDS/AF-6) domain family (N-terminal) member 8, ras association domain-containing protein 8 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction