Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RASSF8 Monoclonal Antibody (OTI3F7)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA526226
Description
RASSF8 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
RASSF8 encodes a member of the Ras-association domain family (RASSF) of tumor suppressor proteins. This gene is essential for maintaining adherens junction function in epithelial cells and has a role in epithelial cell migration. It is a lung tumor suppressor gene candidate. A chromosomal translocation t(12;22)(p11.2;q13.3) leading to the fusion of this gene and the FBLN1 gene is found in a complex type of synpolydactyly. Multiple alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
RASSF8 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8NHQ8 | |
RASSF8 | |
Full length human recombinant protein of RASSF8 produced in HEK293T cell. | |
100 μL | |
Primary | |
Human, Monkey | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
OTI3F7 | |
Unconjugated | |
RASSF8 | |
5133400D11Rik; AA815821; AW123240; C12orf2; Carcinoma-associated protein HOJ-1; carcinoma-associated protein HOJ-1 homolog; EGK_03473; HOJ1; HoJ-1; mHoj-1; Ras association (RalGDS/AF-6) domain family (N-terminal) member 8; Ras association (RalGDS/AF-6) domain family 8; Ras association domain family member 8; ras association domain-containing protein 8; RASSF8; RGD1308820 | |
Mouse | |
Affinity chromatography | |
RUO | |
11228 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction