Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RASSF8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104058
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104058 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104058 Supplier Invitrogen™ Supplier No. PA5104058
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total RASSF8.

RASSF8 encodes a member of the Ras-association domain family (RASSF) of tumor suppressor proteins. This gene is essential for maintaining adherens junction function in epithelial cells and has a role in epithelial cell migration. It is a lung tumor suppressor gene candidate. A chromosomal translocation t(12;22)(p11.2;q13.3) leading to the fusion of this gene and the FBLN1 gene is found in a complex type of synpolydactyly. Multiple alternatively spliced transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RASSF8
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene RASSF8
Gene Accession No. Q8CJ96, Q8NHQ8
Gene Alias 5133400D11Rik; AA815821; AW123240; C12orf2; Carcinoma-associated protein HOJ-1; carcinoma-associated protein HOJ-1 homolog; EGK_03473; HOJ1; HoJ-1; mHoj-1; Ras association (RalGDS/AF-6) domain family (N-terminal) member 8; Ras association (RalGDS/AF-6) domain family 8; Ras association domain family member 8; ras association domain-containing protein 8; RASSF8; RGD1308820
Gene Symbols RASSF8
Host Species Rabbit
Immunogen A synthesized peptide derived from human RASSF8(Accession Q8NHQ8), corresponding to amino acid residues S385-V419.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11228, 312846, 71323
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.