Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RASSF8 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987034
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7034 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7034 Supplier Bioss Supplier No. BS6089R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RASSF8 encodes a member of the Ras-association domain family (RASSF) of tumor suppressor proteins. This gene is essential for maintaining adherens junction function in epithelial cells and has a role in epithelial cell migration. It is a lung tumor suppressor gene candidate. A chromosomal translocation t(12;22)(p11.2;q13.3) leading to the fusion of this gene and the FBLN1 gene is found in a complex type of synpolydactyly. Multiple alternatively spliced transcript variants encoding different isoforms have been found for this gene.

Specifications

Antigen RASSF8
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene RASSF8
Gene Accession No. Q8NHQ8
Gene Alias 5133400D11Rik; AA815821; AW123240; C12orf2; Carcinoma-associated protein HOJ-1; Carcinoma-associated protein HOJ-1 homolog; HOJ1; HoJ-1; mHoj-1; Ras association (RalGDS/AF-6) domain family (N-terminal) member 8; Ras association (RalGDS/AF-6) domain family 8; Ras association domain family member 8; Ras association domain-containing protein 8; RASSF8; RGD1308820
Gene Symbols RASSF8
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human RASSF8.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11228
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.