Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RASSF9 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA550480
Description
The antibody detects endogenous levels of total PAMCI protein.
The protein encoded by this gene localizes to perinuclear endosomes. This protein associates with peptidylglycine alpha-amidating monooxygenase, and may be involved with the trafficking of this enzyme through secretory or endosomal pathways.
Specifications
RASSF9 | |
Polyclonal | |
Unconjugated | |
RASSF9 | |
AW322107; PAM COOH-terminal interactor protein 1; Pamci; PCIP1; P-cip1; Peptidylglycine alpha-amidating monooxygenase COOH-terminal interactor; peptidylglycine alpha-amidating monooxygenase COOH-terminal interactor protein-1; Ras association (RalGDS/AF-6) domain family (N-terminal) member 9; Ras association domain family member 9; ras association domain-containing protein 9; RASSF9 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
9182 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
2.5 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
O75901 | |
RASSF9 | |
Fusion protein corresponding to residues near the C terminal of human Ras association (RalGDS/AF-6) domain family (N-terminal) member 9. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction