Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RASSF9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558878
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558878 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558878 Supplier Invitrogen™ Supplier No. PA558878
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YRILIDKLSA EIEKEVKSVC IDINEDAEGE AASELESSNL ESVKCDLEKS MKAGLKIHSH LSGIQKEIKY SDSLLQMKAK EY Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 82%.

The protein encoded by this gene localizes to perinuclear endosomes. This protein associates with peptidylglycine alpha-amidating monooxygenase, and may be involved with the trafficking of this enzyme through secretory or endosomal pathways.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RASSF9
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RASSF9
Gene Accession No. O75901, Q8K342
Gene Alias AW322107; PAM COOH-terminal interactor protein 1; Pamci; PCIP1; P-cip1; Peptidylglycine alpha-amidating monooxygenase COOH-terminal interactor; peptidylglycine alpha-amidating monooxygenase COOH-terminal interactor protein-1; Ras association (RalGDS/AF-6) domain family (N-terminal) member 9; Ras association domain family member 9; ras association domain-containing protein 9; RASSF9
Gene Symbols RASSF9
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RASSF9. Recombinant protein control fragment (Product #RP-97894).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 237504, 9182
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.