Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat ALK-7 Alexa Fluor 405-conjugated Antibody, R&D Systems™

Catalog No. FAB5771V100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB5771V100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB5771V100 Supplier R&D Systems Supplier No. FAB5771V100UG
This item is not returnable. View return policy

Mouse Monoclonal Antibody

ALK-7/Activin Receptor Type 1C Monoclonal specifically detects ALK-7/Activin Receptor Type 1C in Rat samples. It is validated for Flow Cytometry.

Specifications

Antigen ALK-7/Activin Receptor Type 1C
Applications Flow Cytometry
Classification Monoclonal
Clone 118311
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 405
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. NP_620790
Gene Alias activin A receptor, type IC, Activin receptor type IC, activin receptor type-1C, Activin receptor-like kinase 7, Activin RIC, ACTR-IC, ACVR1C, ACVRLK7, ALK7, ALK7ALK-7, EC 2.7.11, EC 2.7.11.30
Gene Symbols ACVR1C
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant rat ALK-7 Leu26-Arg108 Accession # NP_620790
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 130399
Test Specificity Detects rat ALK-7 in Western blots. In Western blots, no cross-reactivity with recombinant human Activin R1B is observed.
Target Species Rat
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2a
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.