Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rat ALK-7 Alexa Fluor 594-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB5771T100UG
This item is not returnable.
View return policy
Description
ALK-7/Activin Receptor Type 1C Monoclonal specifically detects ALK-7/Activin Receptor Type 1C in Rat samples. It is validated for Flow Cytometry.Specifications
ALK-7/Activin Receptor Type 1C | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
NP_620790 | |
ACVR1C | |
Mouse myeloma cell line NS0-derived recombinant rat ALK-7 Leu26-Arg108 Accession # NP_620790 | |
RUO | |
130399 | |
Rat | |
IgG2a |
Flow Cytometry | |
118311 | |
Alexa Fluor 594 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
activin A receptor, type IC, Activin receptor type IC, activin receptor type-1C, Activin receptor-like kinase 7, Activin RIC, ACTR-IC, ACVR1C, ACVRLK7, ALK7, ALK7ALK-7, EC 2.7.11, EC 2.7.11.30 | |
Mouse | |
100 μg | |
Primary | |
Detects rat ALK-7 in Western blots. In Western blots, no cross-reactivity with recombinant human Activin R1B is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction