Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rat ALK-7 Biotinylated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems BAM5771
This item is not returnable.
View return policy
Description
ALK-7/Activin Receptor Type 1C Monoclonal specifically detects ALK-7/Activin Receptor Type 1C in Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.Specifications
ALK-7/Activin Receptor Type 1C | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 8-25 ug/mL | |
NP_620790 | |
ACVR1C | |
Mouse myeloma cell line NS0-derived recombinant rat ALK-7 Leu26-Arg108 Accession # NP_620790 | |
100 μg | |
Primary | |
Detects rat ALK-7 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human Activin R1B is observed. | |
Rat | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry | |
118311 | |
Biotin | |
Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein. with No Preservative | |
activin A receptor, type IC, Activin receptor type IC, activin receptor type-1C, Activin receptor-like kinase 7, Activin RIC, ACTR-IC, ACVR1C, ACVRLK7, ALK7, ALK7ALK-7, EC 2.7.11, EC 2.7.11.30 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
130399 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction