Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat anti-Abca1, Clone: 5A1-1422.11, Abnova™

Catalog No. 89111000 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-000 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-000 Supplier Abnova Corporation Supplier No. MAB2419
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Specifications

Antigen Abca1
Applications Flow Cytometry
Classification Monoclonal
Clone 5A1-1422.11
Conjugate Unconjugated
Description Rat monoclonal antibody raised against recombinant Abca1.
Dilution Flow Cytometry (1:1000-1:30000) The optimal working dilution should be determined by the end user.
Formulation In antiserum (0.09% sodium azide)
Gene Alias Abc1
Gene Symbols Abca1
Host Species Rat
Immunogen Recombinant protein corresponding extracellular region of mouse Abca1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11303
Test Specificity This antibody is specific to ABCA1.
Target Species Mouse
Content And Storage Store at -20°C or -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.