Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ABCC10, Rat, Clone: M7I-3, Abnova™

Catalog No. 89112997 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-997 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-997 Supplier Abnova Corporation Supplier No. MAB6672
Add to Cart
Add to Cart

Rat monoclonal antibody raised against partial recombinant ABCC10.

The protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, and White). This ABC full-transporter is a member of the MRP subfamily which is involved in multi-drug resistance. Alternative splicing of this gene results in multiple transcript variants; however, not all variants have been fully described. [provided by RefSeq

Specifications

Antigen ABCC10
Applications Immunocytochemistry, Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone M7I-3
Conjugate Unconjugated
Description Rat monoclonal antibody raised against partial recombinant ABCC10.
Dilution Immunohistochemistry (Frozen sections) (1:20) Immunocytochemistry (1:20-1:50) Wstern Blot (1:20-1:50) The optimal working dilution should be determined by the end user.
Formulation In serum-free culture supernatant (0.7% BSA, 0.09% sodium azide)
Gene ABCC10
Gene Alias EST182763/MRP7/SIMRP7
Gene Symbols ABCC10
Host Species Rat
Immunogen Recombinant protein corresponding to amino acids 194-272 of human ABCC10.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 89845
Test Specificity M7I-3 reacts with an internal epitope of MRP7 (ABCC10), an approximately 160 kD transmembrane protein that is related to the multidrug resistance protein MRP1.
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.