Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ABCC12, Rat, Clone: M9I-38, Abnova™

Catalog No. 89112998 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-998 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-998 Supplier Abnova Corporation Supplier No. MAB6674
Add to Cart
Add to Cart

Rat monoclonal antibody raised against partial recombinant ABCC12.

This gene is a member of the superfamily of ATP-binding cassette (ABC) transporters and the encoded protein contains two ATP-binding domains and 12 transmembrane regions. ABC proteins transport various molecules across extra- and intracellular membranes. ABC genes are divided into seven distinct subfamilies: ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, and White. This gene is a member of the MRP subfamily which is involved in multi-drug resistance. This gene and another subfamily member are arranged head-to-tail on chromosome 16q12.1. Increased expression of this gene is associated with breast cancer. [provided by RefSeq

Specifications

Antigen ABCC12
Applications Immunocytochemistry, Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone M9I-38
Conjugate Unconjugated
Description Rat monoclonal antibody raised against partial recombinant ABCC12.
Dilution Immunohistochemistry (Frozen sections) (1:20) Immunocytochemistry (1:20-1:50) Wstern Blot (1:20-1:50) The optimal working dilution should be determined by the end user.
Formulation In serum-free culture supernatant (0.7% BSA, 0.09% sodium azide)
Gene ABCC12
Gene Alias MGC27071/MRP9
Gene Symbols ABCC12
Host Species Rat
Immunogen Recombinant protein corresponding to amino acids 1-42 of human ABCC12.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 94160
Test Specificity M9I-38 reacts with an internal epitope of MRP9 (ABCC12), an approximately 150 kD transmembrane protein that is related to the multidrug resistance protein MRP1.
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.