Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcc5, Rat, Clone: M5I-10, Abnova™

Catalog No. 89112996 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-996 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-996 Supplier Abnova Corporation Supplier No. MAB6671
Add to Cart
Add to Cart

Rat monoclonal antibody raised against partial recombinant Abcc5.

Specifications

Antigen Abcc5
Applications Immunocytochemistry, Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone M5I-10
Conjugate Unconjugated
Description Rat monoclonal antibody raised against partial recombinant Abcc5.
Dilution Immunohistochemistry (Frozen sections) (1:20) Immunocytochemistry (1:20-1:50) Wstern Blot (1:20-1:50) The optimal working dilution should be determined by the end user.
Formulation In serum-free culture supernatant (0.7% BSA, 0.09% sodium azide)
Gene Abcc5
Gene Alias 2900011L11Rik/AI132311/Abcc5a/Abcc5b/MGC116567/Mrp5
Gene Symbols Abcc5
Host Species Rat
Immunogen Recombinant protein corresponding to amino acids 1-38 of mouse Abcc5.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27416
Test Specificity M5I-10 reacts with an internal epitope of MRP5 (ABCC5), an approximately 160 kD transmembrane protein that is related to the multidrug resistance protein MRP1. The monoclonal antibody also strongly reacts with the human MRP5 protein.
Target Species Human, Mouse
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.