Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcg2, Rat, Clone: BXP-9, Abnova™

Catalog No. 89111590 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-590 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-590 Supplier Abnova Corporation Supplier No. MAB3780
Add to Cart
Add to Cart

Rat monoclonal antibody raised against partial recombinant Abcg2.

Specifications

Antigen Abcg2
Applications Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone BXP-9
Conjugate Unconjugated
Description Rat monoclonal antibody raised against partial recombinant Abcg2.
Dilution Immunocytochemistry (1:20-1:50) Immunohistochemistry (Frozen sections) (1:20) Western Blot (1:20-1:50) The optimal working dilution should be determined by the end user.
Formulation In serum-free culture supernatant (0.09% sodium azide)
Gene Abcg2
Gene Alias ABC15/ABCP/AI428558/BCRP/Bcrp1/MXR/MXR1
Gene Symbols Abcg2
Host Species Rat
Immunogen Recombinant MBP fusion protein corresponding to amino acids 221-394 of mouse Abcg2.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26357
Test Specificity BXP-9 reacts with an internal epitope of Breast Cancer Resistance Protein (Bcrp, Abcg2), a 70 KDa transmembrane half-transporter which is involved in multidrug resistance. Does not react with human BCRP.
Target Species Mouse
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.