Learn More
Specifications
Specifications
| Antigen | Aflatoxin |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | IE6 |
| Conjugate | Unconjugated |
| Description | Rat monoclonal antibody raised against Aflatoxin. |
| Dilution | ELISA (5 ng) The optimal working dilution should be determined by the end user. |
| Formulation | Lyophilized from 10mM PBS, pH 7.2 |
| Host Species | Rat |
| Immunogen | Winstar rats were immunized with Aflatoxin M1 conjugated with BSA. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.